Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triosephosphate isomerase Antibody (SR1406), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32218550UL
Description
Triosephosphate isomerase Monoclonal antibody specifically detects Triosephosphate isomerase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
Triosephosphate isomerase | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1406 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
EC 5.3.1.1, MGC88108, TIM, TPI, triosephosphate isomerase, Triose-phosphate isomerase, triosephosphate isomerase 1 | |
A synthesized peptide derived from human Triosephosphate isomerase (Uniprot #: P60174) | |
50 μg | |
Stem Cell Markers | |
7167 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction