Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIP Monoclonal Antibody (2E7)
GREENER_CHOICE

Catalog No. PIMA520999
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520999 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520999 Supplier Invitrogen™ Supplier No. MA520999
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GQSCAGEPDE ELVGAFPIFV RNAILGQKQP KRPRSESSCS KDVVRTGFDG LGGRTKFIQP TDTVMIRPLP VKPKTKVKQR VRVKTVPSLF QAKLDTFLWS.

TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TRAIP
Gene Accession No. Q9BWF2
Gene Alias E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; RING-type E3 ubiquitin transferase TRAIP; RNF206; SCKL9; TRAF interacting protein; TRAF-interacting protein; TRAIP; Trip
Gene Symbols TRAIP
Host Species Mouse
Immunogen TRAIP (NP_005870, 370 a.a. ~ 469 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10293
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.