Learn More
Invitrogen™ TRIP Polyclonal Antibody

Description
Immunogen sequence: LNLPPVASET VDRLVLESPA PVEVNLKLRR PSFRDDIDLN ATFDVDTPPA RPSSSQHGYY EKLCLEKSHS PIQDVPKKIC KGPRKESQLS L Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 60%.
Specifications
Specifications
| Antigen | TRIP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRAIP |
| Gene Accession No. | Q9BWF2 |
| Gene Alias | E3 ubiquitin-protein ligase TRAIP; RING finger protein 206; RING-type E3 ubiquitin transferase TRAIP; RNF206; SCKL9; TRAF interacting protein; TRAF-interacting protein; TRAIP; Trip |
| Gene Symbols | TRAIP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.