Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18487425UL
Description
TRIP1 Polyclonal specifically detects TRIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TRIP1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
eIF-3-beta, eIF3-beta, eIF3i, eIF3-p36, EIF3S2TGF-beta receptor-interacting protein 1, Eukaryotic translation initiation factor 3 subunit 2, eukaryotic translation initiation factor 3 subunit I, eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD), eukaryotic translation initiation factor 3, subunit 2 beta, 36kDa, eukaryotic translation initiation factor 3, subunit I, predicted protein of HQ2242, TGFbeta receptor-interacting protein 1, TRIP-1, TRIP-1eIF3 p36, TRIP1PRO2242 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of human TRIP1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EIF3I | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MGYQCFVSFFDLRDPSQIDNNEPYMKIPCNDSKITSAVWGPLGECIIAGHESGELNQYSAKSGEVLVNVKEHSRQINDIQLSRD | |
25ul | |
Angiogenesis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
8668 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction