Learn More
TRIP1/SUG1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$155.13 - $588.46
Specifications
Antigen | TRIP1/SUG1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1582
![]() |
Bethyl Laboratories, Inc
IHC00151T |
10 μL |
Each for $155.13
|
|
|||||
50-157-1581
![]() |
Bethyl Laboratories, Inc
IHC00151 |
100 μL |
Each for $588.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla,
Human SUG1 (also known as p45, Rpt6 and Cim3) is a component of the 19S regulatory subunit of the 26S proteosome complex. It is one of the AAA ATPas found in the 19S regulatory complex that is thought to participate in the unfolding of ubiquitinated proteins in an ATP-dependent manner. It has also been shown to directly interact with the nucleotide excision repair protein XPB.Specifications
TRIP1/SUG1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
S8, SUG-1, SUG1, TBP10, TRIP1, p45, p45/SUG | |
Between 1 and 50 | |
Primary | |
4° C | |
PSMC5 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P62195, P62196 | |
19184, 5705 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.