Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIP230 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551726
Description
Immunogen sequence: QSLGQVGGSL ASLTGQISNF TKDMLMEGTE EVEAELPDSR TKEIEAIHAI LRSENERLKK LCTDLEEKHE ASEIQIKQQS TSYRNQLQQK EVEISHLKAR QIALQDQLLK LQSAAQSVPS GAGVPATTAS SSFAY Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
The Trip230 gene encodes a protein of 1,978 amino acids with an estimated molecular mass of 230 kDa. The gene is located on chromosome 14q31, which is in a region associated with genetic alterations and thyroid hormone response abnormalities. The Trip230 protein interacts with Rb and thyroid hormone receptor (TR) using two distinct domains. Complex formation with Rb is in a hormone independent manner, but when Trip230 forms a complex with TR, it is in a thyroid hormone-dependent manner. After T3 treatment, Trip230 is phosphorylated and transported to the nucleus, suggesting that phosphorylation may be important for its nuclear translocation.
Specifications
| TRIP230 | |
| Polyclonal | |
| Unconjugated | |
| TRIP11 | |
| ACG1A; CEV14; clonal evolution-related gene on chromosome 14 protein; GMAP210; GMAP-210; golgi-associated microtubule-binding protein 210; Golgi-microtubule-associated protein of 210 kDa; thyroid hormone receptor interactor 11; thyroid receptor-interacting protein 11; TR-interacting protein 11; TRIP11; TRIP-11; Trip230 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15643 | |
| TRIP11 | |
| Recombinant protein corresponding to Human TRIP230. Recombinant protein control fragment (Product #RP-89037). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction