Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIP230 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567119 Supplier Invitrogen™ Supplier No. PA567119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLDDVQKKLMS LANSSEGKVD KVLMRNLFIG HFHTPKNQRH EVLRLMGSIL GVRREEMEQL FHDDQGSVTR WMTGWLGGGS KSVPNTPLRP NQQSV Highest antigen sequence identity to the following orthologs - mouse 91%, rat 92%.

The Trip230 gene encodes a protein of 1,978 amino acids with an estimated molecular mass of 230 kDa. The gene is located on chromosome 14q31, which is in a region associated with genetic alterations and thyroid hormone response abnormalities. The Trip230 protein interacts with Rb and thyroid hormone receptor (TR) using two distinct domains. Complex formation with Rb is in a hormone independent manner, but when Trip230 forms a complex with TR, it is in a thyroid hormone-dependent manner. After T3 treatment, Trip230 is phosphorylated and transported to the nucleus, suggesting that phosphorylation may be important for its nuclear translocation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIP230
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIP11
Gene Accession No. Q15643
Gene Alias ACG1A; CEV14; clonal evolution-related gene on chromosome 14 protein; GMAP210; GMAP-210; golgi-associated microtubule-binding protein 210; Golgi-microtubule-associated protein of 210 kDa; thyroid hormone receptor interactor 11; thyroid receptor-interacting protein 11; TR-interacting protein 11; TRIP11; TRIP-11; Trip230
Gene Symbols TRIP11
Host Species Rabbit
Immunogen Recombinant Human TRIP230. Recombinant protein control fragment (Product #RP-107763).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.