Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIP6 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079791IR
Description
TRIP6 Polyclonal antibody specifically detects TRIP6 in Human samples. It is validated for Western BlotSpecifications
TRIP6 | |
Polyclonal | |
Western Blot | |
MGC10556, MGC10558, MGC29959, MGC3837, MGC4423, OIP1OIP-1, Opa-interacting protein 1, thyroid hormone receptor interactor 6, thyroid receptor-interacting protein 6, TR-interacting protein 6, ZRP-1TRIP-6, zyxin related protein 1, Zyxin-related protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human Thyroid Hormone Receptor Interactor 6 using the numbering given in entry NP_003293.1 (GeneID 7205). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
7205 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction