Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIP6 Monoclonal Antibody (6H4)
GREENER_CHOICE

Catalog No. PIMA517191
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517191 Supplier Invitrogen™ Supplier No. MA517191
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17191 targets TRIP6 in FACS, pep-ELISA, and WB applications and shows reactivity with Human samples. The MA5-17191 immunogen is purified recombinant fragment of human TRIP6 (amino acids: 107-291) expressed in E. Coli. MA5-17191 detects TRIP6 which has a predicted molecular weight of approximately 50.3kDa.

This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIP6
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TRIP6
Gene Accession No. Q15654
Gene Alias MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP1; OIP-1; OPA-interacting protein 1; tcag7.1300; thyroid hormone receptor interacting protein 6; thyroid hormone receptor interactor 6; thyroid receptor-interacting protein 6; TR-interacting protein 6; Trip6; TRIP-6; TRIP6i2; Zrp1; ZRP-1; zyxin related protein 1; zyxin-related protein 1
Gene Symbols TRIP6
Host Species Mouse
Immunogen Purified recombinant fragment of human TRIP6 (amino acids: 107-291) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7205
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.