Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113946 Supplier Invitrogen™ Supplier No. PA5113946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASTPAGPAFP VQVKVAQPVR GCGPPRRGAS QASGPLPGPH FPLPGRGEVW For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIP6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRIP6
Gene Accession No. Q15654
Gene Alias MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP1; OIP-1; OPA-interacting protein 1; tcag7.1300; thyroid hormone receptor interacting protein 6; thyroid hormone receptor interactor 6; thyroid receptor-interacting protein 6; TR-interacting protein 6; Trip6; TRIP-6; TRIP6i2; Zrp1; ZRP-1; zyxin related protein 1; zyxin-related protein 1
Gene Symbols TRIP6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TRIP6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7205
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.