Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tripeptidyl-Peptidase I/TPP1 Rabbit anti-Human, Mouse, Rat, Clone: S09-2G9, Novus Biologicals™

Rabbit Monoclonal Antibody
$274.50 - $586.50
Specifications
Antigen | Tripeptidyl-Peptidase I/TPP1 |
---|---|
Clone | S09-2G9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Tripeptidyl-Peptidase I/TPP1 Monoclonal antibody specifically detects Tripeptidyl-Peptidase I/TPP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Tripeptidyl-Peptidase I/TPP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
1200 | |
IgG | |
Affinity purified |
S09-2G9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2EC 3.4.14.9, growth-inhibiting protein 1, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I | |
Recombinant protein of human Tripeptidyl-Peptidase I/TPP1 (Uniprot#: O14773) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title