Alfa Aesar™ Triphenyl borate, 97%

Manufacturer: Alfa Aesar™ H5568606

Quantity 5g

 View more versions of this product

Catalog No. AAH5568606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1095-03-0
    Linear Formula (C6H5O)3B
    Formula Weight 290.13
    MDL Number MFCD00059011
    UN Number UN2811
    Chemical Name or Material Triphenyl borate

    Specifications

    Formula Weight 290.13
    Melting Point 98° to 101°C
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    CAS 1095-03-0
    UN Number UN2811
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    RTECS Number ED5775000
    Linear Formula (C6H5O)3B
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Sensitivity Moisture Sensitive
    Quantity 5g
    MDL Number MFCD00059011
    Chemical Name or Material Triphenyl borate
    Assay 97%

    Description

    Applications
    Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

    Solubility
    Soluble in chloroform, methanol.

    Notes
    Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.

    GHS H Statement
    H301-H311-H331-H318-H315-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit