Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl phosphate, 99+%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
5kg
Packaging:
Plastic bottle
Plastic drum
115-86-6
C18H15O4P
326.28
MFCD00003031
XZZNDPSIHUTMOC-UHFFFAOYSA-N
triphenylphosphate,phosphoric acid, triphenyl ester,disflamoll tp,triphenoxyphosphine oxide,celluflex tpp,phosflex tpp,trifenylfosfat,phenyl phosphate pho 3po,phosphoric acid triphenyl ester,trifenylfosfat czech
8289
CHEBI:35033
triphenyl phosphate
C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)OC3=CC=CC=C3
115-86-6
C18H15O4P
326.28
MFCD00003031
XZZNDPSIHUTMOC-UHFFFAOYSA-N
triphenylphosphate,phosphoric acid, triphenyl ester,disflamoll tp,triphenoxyphosphine oxide,celluflex tpp,phosflex tpp,trifenylfosfat,phenyl phosphate pho 3po,phosphoric acid triphenyl ester,trifenylfosfat czech
8289
CHEBI:35033
triphenyl phosphate
C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)OC3=CC=CC=C3
Boiling Point 370°C
Flash Point 220°C
Quantity 5kg
Infrared Spectrum Authentic
Acidity 0.05mg KOH/g max.
Packaging Plastic drum
Color White
Melting Point 47°C to 53°C
Water 0.1% max.
Assay Percent Range 99+%
Linear Formula (C6H5O)3P(O)
Beilstein 06, 179
Merck Index 15, 9915
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and acetone
Formula Weight 326.28
Physical Form Crystalline Flakes
Percent Purity ≥99.0%
Chemical Name or Material Triphenyl phosphate
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning