Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triphenyl Phosphate Solution, NMR Reference Standard, 0.03357 M in chloroform-d (99.8 atom % D), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
0.7 mL
Specifications
Specifications
| Packaging | NMR Tube Size: 5 mm 8 in. |
| Concentration | 0.0336 M in chloroform-d (99.8 atom% D) |
| Linear Formula | (C6H5O)3 PO |
| MDL Number | MFCD00003031 |
| Quantity | 0.7 mL |
| Formula Weight | 326.28 |
| Grade | NMR Reference Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction