Learn More
Triphenyl phosphite, 97%
Description
Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used to convert alcohols to alkyl halides, as a peptide coupling agent, and, in combination with ozone, as a low-temperature source of singlet oxygen, and ligand for metal-catalyzed reactions. Also used as reagent in the preparation of mixed valent nickel diphosphine dithiolate bridged iron carbonyl phosphine complexes as models of [NiFe]-hydrogenase active site.
Solubility
Insoluble in water.
Notes
Air and Moisture Sensitive, store away form air and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Specifications
Specifications
| Melting Point | 22°C to 24°C |
| Density | 1.184 |
| Boiling Point | 360°C |
| Flash Point | 146°C (295°F) |
| Odor | Mild |
| Linear Formula | (C6H5O)3P |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H317-H319 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 202-908-4 |
| RTECSNumber | TH1575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.