Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl phosphite, 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
101-02-0
C18H15O3P
310.28
HVLLSGMXQDNUAL-UHFFFAOYSA-N
phosphorous acid, triphenyl ester,advance tpp,triphenoxyphosphine,phenyl phosphite,stabilizer p 36,trifenoxyfosfin,trifenylfosfit,phosclere t 36,efed,mellite 310
7540
triphenyl phosphite
C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3
101-02-0
C18H15O3P
310.28
HVLLSGMXQDNUAL-UHFFFAOYSA-N
phosphorous acid, triphenyl ester,advance tpp,triphenoxyphosphine,phenyl phosphite,stabilizer p 36,trifenoxyfosfin,trifenylfosfit,phosclere t 36,efed,mellite 310
7540
triphenyl phosphite
C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3
Acidity 1.5mg KOH/g max.
Color Colorless to Yellow
Boiling Point 360.0°C
Quantity 2.5L
Packaging Glass Bottle
Specific Gravity 1.184
Refractive Index 1.5893 to 1.5913
Infrared Spectrum Authentic
Melting Point 22.0°C to 24.0°C
Flash Point 146°C
Density 1.1840g/mL
Assay Percent Range 99%
Linear Formula (C6H5O)3P
Beilstein 06,177
Fieser 01,1249; 02,446; 03,322
Solubility Information Solubility in water: insoluble. Other solubilities: slowly hydrolyses in water, soluble in many organic solvents
Formula Weight 310.28
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Triphenyl phosphite
Show More Show Less