Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylacetic acid, 99%

Catalog No. p-208660
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle

CAS: 595-91-5 | C20H16O2 | 288.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-91-5
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004185
InChI Key DCYGAPKNVCQNOE-UHFFFAOYSA-N
Synonym triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
PubChem CID 68992
IUPAC Name 2,2,2-triphenylacetic acid
SMILES OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 268°C to 269°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5)3CCO2H
Quantity 5 g
Beilstein 09, 712
Formula Weight 288.34
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Triphenylacetic acid, 99%
Show More Show Less
Hazard Statement GHS H Statement
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
EINECSNumber 209-873-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.