Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylamine, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
603-34-9
C18H15N
245.32
MFCD00003020
ODHXBMXNKOYIBV-UHFFFAOYSA-N
triphenylamine,benzenamine, n,n-diphenyl,triphenyl amine,n,n-diphenylbenzenamine,amine, triphenyl,unii-njs65m2ds2,ccris 4887,n,n,n-triphenylamine,n,n-diphenylbenzeneamine,njs65m2ds2
11775
N,N-diphenylaniline
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3
603-34-9
C18H15N
245.32
MFCD00003020
ODHXBMXNKOYIBV-UHFFFAOYSA-N
triphenylamine,benzenamine, n,n-diphenyl,triphenyl amine,n,n-diphenylbenzenamine,amine, triphenyl,unii-njs65m2ds2,ccris 4887,n,n,n-triphenylamine,n,n-diphenylbenzeneamine,njs65m2ds2
11775
N,N-diphenylaniline
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3
Boiling Point 347°C to 348°C
Quantity 25g
Infrared Spectrum Authentic
Melting Point 125°C to 127°C
Packaging Glass bottle
Color White to Beige
Assay Percent Range 99+%
Linear Formula (C6H5)3N
Beilstein 12, 181
Solubility Information Solubility in water: insoluble
Formula Weight 245.32
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material Triphenylamine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.