Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylborane, 96%

Catalog No. p-7049984
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 960-71-4 | C18H15B | 242.128 g/mol

Triphenylborane is used as a precursor in the production of pyridine-triphenylborane complex, which is used as a catalyst for the polymerization of acrylic esters. It is also used in the hydrocyanation of butadiene to adiponitrile. It is involved in the synthesis of 3-phenyl-cyclohexene by reacting with acetic acid cyclohex-2-enyl ester in presence of palladium bis(dibenzylideneacetone) and triphenylphosphine as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 960-71-4
Molecular Formula C18H15B
Molecular Weight (g/mol) 242.128
MDL Number MFCD00003007
InChI Key MXSVLWZRHLXFKH-UHFFFAOYSA-N
Synonym triphenylboron, borane, triphenyl, triphenyboron, triphenylborine, borine, triphenyl 6ci, boron triphenyl, triphenyl boron, triphenyl-boran, borine,triphenyl, triphenyl-borane
PubChem CID 70400
IUPAC Name triphenylborane
SMILES B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 147°C
Boiling Point 208°C (14 mmHg)
Odor Odorless
Quantity 1 g
UN Number UN1325
Beilstein 1961313
Sensitivity Air Sensitive
Solubility Information Soluble in aromatic solvents. Insoluble in water.
Formula Weight 242.13
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Triphenylborane
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H318-H228
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 213-504-2
RTECSNumber ED2367500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.