Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylborane - Triphenylphosphine Complex 97.0+%, TCI America™
SDP

Catalog No. T175725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T175725G 25 g
T17575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T175725G Supplier TCI America Supplier No. T175725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3053-68-7
Molecular Formula C36H30BP
Molecular Weight (g/mol) 504.42
MDL Number MFCD01631317
InChI Key QLAGHGSFXJZWKY-UHFFFAOYSA-N
PubChem CID 11081676
IUPAC Name triphenylborane;triphenylphosphane
SMILES B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 504.42
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylborane - Triphenylphosphine Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.