Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylcarbenium hexachloroantimonate, 98%, Thermo Scientific™

Catalog No. AC172930250
Click to view available options
Quantity:
25 g
50 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1586-91-0
Molecular Formula C19H15Cl6Sb
Molecular Weight (g/mol) 577.79
MDL Number MFCD00013126
InChI Key GASWLBDVMZXHOP-UHFFFAOYSA-H
Synonym tritylium hexachloroantimonate, triphenylcarbenium hexachloroantimonate, trityl hexachloroantimonate, hexachlorostibanuide; triphenylmethylium
PubChem CID 16688166
IUPAC Name hexachlorostibanuide; triphenylmethylium
SMILES Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (C6H5)3CSbCl6
Quantity 25 g
Formula Weight 577.79
Physical Form Powder
Chemical Name or Material Triphenylcarbenium hexachloroantimonate, 98%
Hazard Category MOISTURE SENSITIVE
EINECSNumber 216-446-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.