Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triphenylcarbenium tetrafluoroborate, 97%
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 205°C to 215°C |
| Color | Green to Ochre-Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% |
| Packaging | Plastic bottle |
| Molecular Formula | C19H15·BF4 |
| Linear Formula | (C6H5)3CBF4 |
| Quantity | 100 g |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in methanol |
| Molecular Weight (g/mol) | 330.13 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Reacts violently with water. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
| EINECSNumber | 206-433-3 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction