Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylchlorosilane 95.0+%, TCI America™
SDP

Catalog No. T093925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T093925G 25 g
T09395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T093925G Supplier TCI America Supplier No. T093925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76-86-8
Molecular Formula C18H15ClSi
Molecular Weight (g/mol) 294.85
MDL Number MFCD00000496
InChI Key MNKYQPOFRKPUAE-UHFFFAOYSA-N
Synonym triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl
PubChem CID 6458
IUPAC Name chlorotriphenylsilane
SMILES Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C
Color White
Boiling Point 208°C
Quantity 25 g
UN Number 3261
Formula Weight 294.85
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylchlorosilane
EINECSNumber (3)-2634
RTECSNumber VV2720000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.