Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylmethanesulfenyl Chloride 96.0+%, TCI America™
SDP

Catalog No. T123710G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 24165-03-5
Molecular Formula C19H15ClS
Molecular Weight (g/mol) 310.84
MDL Number MFCD00010360
InChI Key ZTQHJLXIUQQVST-UHFFFAOYSA-N
Synonym Tritylsulfenyl Chloride
PubChem CID 342378
IUPAC Name [(chlorosulfanyl)diphenylmethyl]benzene
SMILES ClSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 137°C
Color Yellow
Quantity 10 g
UN Number 1759
Formula Weight 310.84
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethanesulfenyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.