Alfa Aesar™ Triphenylmethanol, 98%

Manufacturer: Alfa Aesar™ A1036630

Quantity 250g

 View more versions of this product

Catalog No. AAA1036630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76-84-6
    Molecular Formula (C6H5)3COH
    Formula Weight 260.34
    MDL Number MFCD00004445
    Beilstein 1460837
    Merck Index 14,9739
    Synonym Triphenyl carbinol; Trityl alcohol
    Chemical Name or Material Triphenylmethanol

    Specifications

    Formula Weight 260.34
    Assay Percent Range 98%
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 76-84-6
    TSCA TSCA
    MDL Number MFCD00004445
    Merck Index 14,9739
    Health Hazard 1 Warning
    Melting Point 161° to 164°C
    Beilstein 1460837
    Synonym Triphenyl carbinol; Trityl alcohol
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula (C6H5)3COH
    Boiling Point 360° to 380°C
    Quantity 250g
    Chemical Name or Material Triphenylmethanol
    Assay 98%

    Description

    Applications
    Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

    Solubility
    Soluble in dioxane, ethanol, water, ether and benzene.

    Notes
    Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit