Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine, 98%, Spectrum™ Chemical
SDP

Catalog No. 18614571
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
50 lb.
220 lb.
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 603-35-0

Specifications

CAS Max % 1
Boiling Point 360°C
Packaging Amber Glass Bottle
Quantity 220 lb.
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.