Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine, 99+%

Catalog No. p-4408501
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 603-35-0 | C18H15P | 262.29 g/mol

Triphenylphosphine is used in the synthesis of organic compounds due to its nucleophilicity and its reducing character. It is involved in the synthesis of biaryl compounds, phosphonium salts and other phosphorus compounds. As a reducing agent, it is used to prepare aromatic amines from the corresponding aromatic N-oxides. The anionic phosphine is usually isolated as the trisodium salt which reacts with rhodium to form a complex that finds use in industrial hydroformylation reactions. It is also used to prepare Wilkinson's catalyst, RhCl(PPh3)3 useful to catalyze the hydrogenation of alkenes and tetrakis(triphenylphosphine)palladium(0) that is widely used to catalyze C-C coupling reactions in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-35-0
Molecular Formula C18H15P
Molecular Weight (g/mol) 262.29
MDL Number MFCD00003043 MFCD20489348
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
Synonym triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin
PubChem CID 11776
IUPAC Name triphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 79°C to 81°C
Density 1.2 g/mL
Boiling Point 360°C
Flash Point 181°C (358°F)
Assay Percent Range >99%
Quantity 25 g
Beilstein 610776
Solubility Information Soluble in ether,benzene,carbon tetrachloride,glacial acetic acid,acetone,chloroform and alcohol. Insoluble in water.
Formula Weight 262.29
Physical Form Flakes
Chemical Name or Material Triphenylphosphine
Show More Show Less
Hazard Category H302-H317-H373
Precautionary Statement P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
EINECSNumber 210-036-0
RTECSNumber SZ3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.