Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine resin, 1% crossl. with DVB, 1.0-1.5 mmol/g, 100-200 mesh

Catalog No. p-2692413
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 39319-11-4 | C18H15P | 262.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39319-11-4
Molecular Formula C18H15P
Molecular Weight (g/mol) 262.29
MDL Number MFCD00003043 MFCD20489348
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
PubChem CID 11776
IUPAC Name triphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Physical Form 100 To 200 Mesh
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.