Learn More
Triphenylphosphine sulfide, 98%, Thermo Scientific Chemicals
Description
Triphenylphosphine sulfide involves in the , conversion of epoxides to episulfides (thiiranes) in presence of TFA of the same stereochemistry. It acts as a ligand for the Pd-catalyzed bisalkoxycarbonylation of olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triphenylphosphine sulfide involves in the , conversion of epoxides to episulfides (thiiranes) in presence of TFA of the same stereochemistry. It acts as a ligand for the Pd-catalyzed bisalkoxycarbonylation of olefins.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 158°C to 164°C |
| Quantity | 100 g |
| UN Number | UN3464 |
| Beilstein | 645872 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 294.36 |
| Percent Purity | 98% |
| Chemical Name or Material | Triphenylphosphine sulfide |
Safety and Handling
| Hazard Category | H302+H312 |
| Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S. |
| EINECSNumber | 223-407-7 |
| RTECSNumber | SZ2820000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.