Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyltin chloride, 95%

Catalog No. p-7046425
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 639-58-7 | C18H17ClSn | 387.49 g/mol

Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (0.005g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 639-58-7
Molecular Formula C18H17ClSn
Molecular Weight (g/mol) 387.49
MDL Number MFCD00000519
InChI Key ZDYYCRIXXMWWQC-UHFFFAOYSA-N
Synonym triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
PubChem CID 12540
ChEBI CHEBI:35036
IUPAC Name chloro(triphenyl)stannane
SMILES Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 103°C to 106°C
Boiling Point 240°C (13.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Quantity 25 g
UN Number UN3146
Beilstein 524762
Sensitivity Moisture sensitive
Solubility Information Soluble in water (0.005g/L at 20°C).
Formula Weight 385.48
Percent Purity 95%
Chemical Name or Material Triphenyltin chloride
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 211-358-4
RTECSNumber WH6860000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.