Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylvinylsilane 93.0+%, TCI America™
SDP

Catalog No. T169525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T169525G 25 g
T16955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T169525G Supplier TCI America Supplier No. T169525G

Chemical Identifiers

CAS 18666-68-7
Molecular Formula C20H18Si
Molecular Weight (g/mol) 286.449
MDL Number MFCD00051542
InChI Key OVOIHGSHJGMSMZ-UHFFFAOYSA-N
Synonym triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane
PubChem CID 87747
IUPAC Name ethenyl(triphenyl)silane
SMILES C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 69°C
Color White-Yellow
Boiling Point 190°C
Quantity 25 g
Formula Weight 286.45
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylvinylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.