Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Catalog No. 365H2181
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
Needle Gauge:
23 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Needle Gauge
365H2181 100 μL 23 G
365I2351 250 μL 23 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 365H2181 Supplier Thermo Scientific™ Supplier No. 365H2181
Add to Cart
Add to Cart
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Needle), Stainless Steel (Plunger)
Description 100 μL FN GT, 23 G, 57 mm
Needle Gauge 23 G
Length (Metric) 57 mm
Product Type GC Autosampler Syringe
Quantity Ea.
Volume (Metric) 100 μL
Product Line TriPlus
Length (Metric) Needle 57 mm
Needle Point Style Side Hole
For Use With (Equipment) TriPlus RSH Autosampler
Connection Type Fixed
PTFE Tipped Plunger Yes