Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropargylamine, 97%

Catalog No. aah5345903
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 6921-29-5 | C9H9N | 131.178 g/mol

Tripropargylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripropargylamine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6921-29-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00008577
InChI Key ZHOBJWVNWMQMLF-UHFFFAOYSA-N
Synonym tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb
PubChem CID 23351
IUPAC Name N,N-bis(prop-2-ynyl)prop-2-yn-1-amine
SMILES C#CCN(CC#C)CC#C

Specifications

Density 0.927
Boiling Point 79°C to 85°C (11 mmHg)
Flash Point 57°C (135°F)
Refractive Index 1.484
Linear Formula (HC≡CCH2)3N
Quantity 1 g
UN Number UN1993
Formula Weight 131.18
Percent Purity 97%
Chemical Name or Material Tripropargylamine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 230-038-5
RTECSNumber YK7660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.