Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tripropylene glycol

Manufacturer:  Alfa Aesar™ 31798AK

Quantity 250mL

 View more versions of this product

Catalog No. AA31798AK


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

24800-44-0
192.25
Tripropylene glycol
HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH
MFCD00014405

Specifications

24800-44-0
HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH
MFCD00014405
1.018g/mL
Odorless
Tripropylene glycol
Liquid
192.25
1.4443
268°C
250mL

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit