Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropylene glycol

Catalog No. AA31798K2
Click to view available options
Quantity:
250 mL
1 L
4 L

Catalog No. AA31798K2

Supplier: Thermo Scientific Chemicals 031798K2

Only null left
Add to Cart

CAS: 24800-44-0 | HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH

Tripropylene glycol is used for textile soaps, lubricants, cutting oils and greases. It acts as an intermediate in the preparation of urethanes. It is also used as a solvent in coatings and inks. It is utilized as varnish removers, emulsifiers and hydraulic fluids. As an additive, it is used for carburetor fuels. It is involved as a cooling media as well as an active ingredient of plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24800-44-0
Molecular Formula HOCH(CH3)CH2OCH2CH(CH3)OCH2CH(CH3)OH
MDL Number MFCD00014405

Specifications

Density 1.018 g/mL
Boiling Point 268°C
Refractive Index 1.4443
Quantity 1 L
Solubility Information Miscible with water,methanol and ether.
Formula Weight 192.25
Physical Form Liquid
Chemical Name or Material Tripropylene glycol
EINECSNumber 246-466-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.