Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropylene Glycol (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. p-7136326
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 24800-44-0
Molecular Formula C9H20O4
MDL Number MFCD00014405

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripropylene Glycol (mixture of isomers)
EINECSNumber (2)-0430
RTECSNumber YK6825000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.