Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Triptolide
GREENER_CHOICESDP

Catalog No. 325310 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-531-0 10 mg
32-531 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-531-0 Supplier R&D Systems™ Supplier No. 3253/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits RNAPII-mediated transcription; antitumor, anti-inflammatory and immunosuppressive

Triptolide inhibits DNA-dependent ATPase activity of XBP and induces inhibition of RNA polymerase II (RNAPII)-mediated transcription (IC50 = 200 nM). Selective for RNAPII over RNAPI and RNAPIII. Triptolide blocks RNA synthesis in HeLa cells (IC50 = 62 nM); exhibits potent antiproliferative activity in 60 cancer cell lines (average IC50 = 12 nM) and induces apoptosis by blocking TNF-α-mediated c-IAP1 and c-IAP2 induction. Also displays immunosuppressive and anti-inflammatory activity.

Chemical Identifiers

CAS 38748-32-2
Molecular Formula C20H24O6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 360.4
Purity >98%
Molecular Weight (g/mol) 360.41
Chemical Name or Material Triptolide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.