Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIQK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585011 Supplier Invitrogen™ Supplier No. PA585011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFFYLRLTTD VDPDLDQDED Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 75%.

TRIQK is a protein coding gene. May play a role in cell growth and maintenance of cell morphology. May play a role in cell growth and maintenance of cell morphology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIQK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIQK
Gene Accession No. Q629K1
Gene Alias C8orf83; DKFZp779L1068; LOC286144; PRO0845; protein TRIQK; triple QxxK/R motif containing; triple QxxK/R motif-containing protein; Triple repetitive-sequence of QXXK/R protein homolog; TRIQK; UPF0599
Gene Symbols TRIQK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIQK. Recombinant protein control fragment (Product #RP-108671).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286144
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.