Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triquinoyl Hydrate 95.0+%, TCI America™
SDP

Catalog No. T08761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08761G 1 g
T08765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08761G Supplier TCI America Supplier No. T08761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 527-31-1
Molecular Formula C6O6
Molecular Weight (g/mol) 168.06
MDL Number MFCD00001652,MFCD00149074
InChI Key PKRGYJHUXHCUCN-UHFFFAOYSA-N
Synonym cyclohexanehexone, triquinoyl, cyclohexane-1,2,3,4,5,6-hexaone, cyclohexanehexaone, hexaketocyclohexane, unii-7zr8062lfd, trichinoyl, hexaoxocyclohexane, hexaoxocyclohexaneoctahydrate, cyclohexane-hexone, octahydrate
PubChem CID 68240
IUPAC Name cyclohexane-1,2,3,4,5,6-hexone
SMILES O=C1C(=O)C(=O)C(=O)C(=O)C1=O

Specifications

Melting Point 93°C
Quantity 1 g
Formula Weight 168.06
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triquinoyl Hydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.