Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS, 0.5 M Buffer Solution, pH 6.8, PROMO, Thermo Scientific Chemicals

Catalog No. J63735APPM
Click to view available options
Quantity:
500 mL
1 L

Applications

TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.

Solubility

Difficult to mix.

Notes

Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14 g/mol
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 0.5 M
pH 6.8
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material TRIS buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.