Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%

Catalog No. AA4120203
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4120203 1 g
AA41202MD 250 mg
AA4120206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4120203 Supplier Thermo Scientific Chemicals Supplier No. 04120203
Add to Cart

CAS: 142617-53-6 | C33H57BiO6 | 758.793 g/mol

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142617-53-6
Molecular Formula C33H57BiO6
Molecular Weight (g/mol) 758.793
MDL Number MFCD00064763
InChI Key ZNHBPQSSVCRFST-LWTKGLMZSA-K
Synonym tris 2,2,6,6-tetramethyl-3,5-heptanedionato bismuth iii, bismuth iii tetramethylheptanedionate, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy bismuthanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
PubChem CID 16717622
IUPAC Name (Z)-5-bis[[(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl]oxy]bismuthanyloxy-2,2,6,6-tetramethylhept-4-en-3-one
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O[Bi](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 114°C to 116°C
Color White to Brown
Boiling Point 150°C (0.05 mmHg)
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 758.79
Percent Purity 99.9%
Physical Form Crystalline
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III)
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.