Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,4-pentanedionato)chromium(III) 98.0+%, TCI America™
SDP

Catalog No. C0336500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0336500G 500 g
C033625G 25 g
C0336100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0336500G Supplier TCI America Supplier No. C0336500G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 21679-31-2
Molecular Formula C15H21CrO6
Molecular Weight (g/mol) 349.32
MDL Number MFCD00000015 MFCD00000015
InChI Key JWORPXLMBPOPPU-LNTINUHCSA-K
Synonym chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
PubChem CID 91759531
IUPAC Name chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 216°C
Color Purple
Quantity 500 g
Formula Weight 349.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)chromium(III)
EINECSNumber (2)-2142
RTECSNumber GB7575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.