Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2,4-pentanedionato)cobalt(III) 98.0+%, TCI America™
SDP

Catalog No. C0374100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0374100G 100 g
C037425G 25 g
C0374500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0374100G Supplier TCI America Supplier No. C0374100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21679-46-9
Molecular Formula C15H21CoO6
Molecular Weight (g/mol) 356.26
MDL Number MFCD00013488
InChI Key VACIXPOMJJIDBM-UHFFFAOYSA-N
Synonym Acetylacetone Cobalt(III) Salt, Cobalt(III) Acetylacetonate
IUPAC Name 2,4,8,10,13,15-hexamethyl-1λ³,5λ³,7λ³,11λ³,12λ³,16λ³-hexaoxa-6-cobaltaspiro[5.5⁶.5⁶]hexadeca-1,4,7,10,12,15-hexaene-6,6,6-tris(ylium)-3,9,14-triide
SMILES CC1=[O][Co+3]234([O]=C(C)[CH-]1)[O]=C(C)[CH-]C(C)=[O]2.CC([CH-]C(C)=[O]3)=[O]4

Specifications

Quantity 100 g
Formula Weight 356.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)cobalt(III)
EINECSNumber (2)-2142
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.