Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-aminoethyl)amine, 98+%

Catalog No. p-4510543
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

CAS: 4097-89-6 | C6H18N4 | 146.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-89-6
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.24
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
PubChem CID 77731
ChEBI CHEBI:30631
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
SMILES C(CN(CCN)CCN)N

Specifications

Melting Point -16.0°C
Color Yellow
Density 0.9770g/mL
Boiling Point 265.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Linear Formula (H2NCH2CH2)3N
Quantity 100 mL
Beilstein 04,II,695
Specific Gravity 0.977
Solubility Information Solubility in water: soluble.
Formula Weight 146.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tris(2-aminoethyl)amine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 223-857-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.