Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-aminoethyl)amine 98.0+%, TCI America™
SDP

Catalog No. T124325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T124325ML 25 mL
T12435ML 5 mL
T1243100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T124325ML Supplier TCI America Supplier No. T124325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4097-89-6
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.238
MDL Number MFCD00008177
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
PubChem CID 77731
ChEBI CHEBI:30631
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
SMILES C(CN(CCN)CCN)N

Specifications

Color Yellow
Boiling Point 114°C
Quantity 25 mL
UN Number 2922
Formula Weight 146.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tris(2-aminoethyl)amine
RTECSNumber KH8587082
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.