Learn More
Tris(2-cyanoethyl)phosphine, 94%
Description
Tris(2-cyanoethyl)phosphine is used to produce 3,3',3″-phosphanetriyl-tri-propionic acid and hydrochloride by heating. It will need reagent conc. aq. HCl with reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 97°C to 98°C |
| Boiling Point | 235°C (0.9 mmHg) |
| Quantity | 1 g |
| UN Number | UN3464 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in DMSO,and DMF. Insoluble in water. |
| Formula Weight | 193.19 |
| Percent Purity | 94% |
| Physical Form | Crystalline |
| Chemical Name or Material | Tris(2-cyanoethyl)phosphine |
Safety and Handling
| Hazard Category | H302-H315-H319-H331-H335-H350 |
| Hazard Statement | GHS H Statement H300-H330-H315-H319-H335 Fatal if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S. |
| EINECSNumber | 223-687-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.