Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-ethylhexyl) Phosphite 90.0+%, TCI America™
SDP

Catalog No. p-7134986
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 301-13-3
Molecular Formula C24H51O3P
Molecular Weight (g/mol) 418.643
MDL Number MFCD00015266
InChI Key ILLOBGFGKYTZRO-UHFFFAOYSA-N
Synonym Phosphorous Acid Trioctyl Ester, Phosphorous Acid Tris(2-ethylhexyl) Ester, Trioctyl Phosphite
PubChem CID 9320
IUPAC Name tris(2-ethylhexyl) phosphite
SMILES CCCCC(CC)COP(OCC(CC)CCCC)OCC(CC)CCCC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 418.64
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-ethylhexyl) Phosphite
RTECSNumber TH2090000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.