Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris[3-(heptafluoropropylhydroxymethylene)-d-camphorato], ytterbium(III) derivative, 98%, Thermo Scientific™

Catalog No. AC217610010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80464-74-0
Molecular Formula C42H42F21O6Yb
Molecular Weight (g/mol) 1214.812
MDL Number MFCD00151061
InChI Key CGQGCFJFDNPWAI-UHFFFAOYSA-N
Synonym Ytterbium tris[3-heptafluoropropylhydroxymethylene)-(-)-camphorate]
PubChem CID 131885416
SMILES CC1(C2CCC1(C(=O)[C-]2C(=O)C(C(C(F)(F)F)(F)F)(F)F)C)C.CC1(C2CCC1(C(=O)[C-]2C(=O)C(C(C(F)(F)F)(F)F)(F)F)C)C.CC1(C2CCC1(C(=O)[C-]2C(=O)C(C(C(F)(F)F)(F)F)(F)F)C)C.[Yb+3]

Specifications

Melting Point 133°C to 139°C
Color Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition + 124.00 (20.00°C c=1,CH2Cl2)
Specific Rotation +122° to +126° (20°C, 589nm) (c=1, CH2Cl2)
Formula Weight 1214.8
Physical Form Crystalline Powder
Chemical Name or Material Tris[3-(heptafluoropropylhydroxymethylene)-d-camphorato], ytterbium(III) derivative
Show More Show Less
Hazard Category HYGROSCOPIC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.