Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris, ≥98%

Catalog No. p-7082838
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)amino methane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 171°C to 172°C
Boiling Point 219°C to 220°C (10 mmHg)
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula NH2C(CH2OH)3
Packaging Plastic bottle
Beilstein 741883
Formula Weight 121.14
Percent Purity ≥98%
Chemical Name or Material Tris
EINECSNumber 201-064-4
RTECSNumber TY2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.