Learn More
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
Shop All Fisherbrand Products

Description
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TBE buffer solution - simply dilute as needed
- 10X solution contains 0.89M tris, 0.89M borate, and 0.02M EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBE Solution with 9 volumes of ultrapure water, such as BP2819
- Tested for the absence of DNase, RNase, and protease
- Functionally tested for use in electrophoresis applications
- TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
- Intended for laboratory research use only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Colorless |
| pH | 8.2 to 8.4 |
| Physical Form | Liquid |
| Quantity | 1 L |
| DNase | DNase free |
| Filtered Through | Filtered through a 0.2-micron filter. |
| Packaging | Nalgene Poly Bottle |
| Protease | Protease free |
| Grade | DNase-, RNase- and Protease-Free |
| Chemical Name or Material | Tris-Borate-EDTA, 10X Solution |
Safety and Handling
| Recommended Storage | RT |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.